Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klotho Antibody, Novus Biologicals™
SDP

Catalog No. NBP136969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP136969 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP136969 Supplier Novus Biologicals Supplier No. NBP136969
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Klotho Polyclonal specifically detects Klotho in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen Klotho
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004786.2
Gene Alias EC 3.2.1, EC 3.2.1.31, klotho
Gene Symbols KL
Host Species Goat
Immunogen Peptide with sequence NQSQVNHTILQYYRC corresponding to internal region according to NP_004786.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9365
Test Specificity This antibody recognizes and epitope within the internal region.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.