Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klotho beta Antibody (1025C), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB377398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377398 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377398 Supplier Novus Biologicals Supplier No. FAB58891A
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Klotho beta Monoclonal specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Klotho beta
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1025C
Conjugate APC
Dilution Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias beta-klotho, betaKlotho, BKL, klotho beta, klotho beta like, klotho beta-like protein, MGC142213
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived recombinant human Klotho beta with a C-terminal 10 His tag., Phe53-Leu997, Accession # Q86Z14
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152831
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.