Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Klotho, Rat anti-Mouse, Human, Clone: KL-234 , Millipore Sigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABN1807100UL
Description
Anti-Klotho Antibody, clone KL-234, Cat. No. MABN1807, is a highly specific rat monoclonal antibody that targets Klotho and has been tested for use in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Klotho (UniProt: Q9UEF7; also known as EC: 3.2.1.31) is encoded by the KL gene (Gene ID:9365) in human. Klotho is a homodimeric, highly N-glycosylated, single-pass type I membrane protein that is present in kidney, placenta, small intestine, and prostate tissue. In kidney it is predominantly present in cortical renal tubules. A soluble secreted form (isoform 2, ∽ 64 kDa) is present in serum and cerebrospinal fluid. The Klotho peptide generated by cleavage of the membrane-bound isoform (Isoform 1) is considered as an anti-aging circulating hormone that is reported to extend life span by inhibiting insulin/IGF1 signaling. It also acts as a cofactor for interaction of FGF23 with FGF R1, which negatively regulates the activity of 1-alpha-hydroxylase, the rate limiting enzyme in the synthesis of 1,25(OH)2D3. Klotho is synthesized with a signal peptide (aa 1-33), which is subsequently cleaved off to generate the mature form that has an extracellular domain (aa 31-981), a short transmembrane domain (aa 982-1002), and a short cytoplasmic tail (aa 1003-1012). Deficiency of Klotho is reported to shorten life-span in animal models with a myriad of disorders associated with old age, such as arteriosclerosis, osteoporosis, and skin atrophy. Klotho deficient mice also display severe hyperphosphatemia and soft tissue calcification. On the contrary, over-expression of Klotho is shown to extend life-span in murine models by about 30% with side effects, such as mild insulin resistance and enhanced resistance to oxidative stress. Defects in KL gene has been linked to chronic renal failure complications. 
            Specifications
| Klotho | |
| Monoclonal | |
| Unconjugated | |
| Rat monoclonal antibody as ascites with 0.1% sodium azide with 50% glycerol. | |
| KL | |
| GST-tagged recombinant fragment corresponding to 107 amino acids from the extracellular domain from the N-terminal region of human Klotho. | |
| 100 μL | |
| Neuroscience | |
| NP_004786 | |
| Mouse, Human | |
| Ascites | 
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| KL-234 | |
| Western Blotting Analysis: A 1:1,000 dilution from a represetative lot detected Klotho in 10μg of mouse kidney tissue lysates.Immunohistochemistry Analysis: A representative lot detected Klotho in Immunohistochemistry applications (Maltare, A., et. al. (2014). Monoclon Atib Immunodiagn Immunother. 33(6):420-7).Immunoprecipitation Analysis: A representative lot immunoprecipitated Klotho in Immunoprecipitation applications (Maltare, A., et. al. (2014). Monoclon Atib Immunodiagn Immunother. 33(6):420-7).Western Blotting Analysis: A representative lot detected Klotho in Western Blotting applications (Maltare, A., et. al. (2014). Monoclon Atib Immunodiagn Immunother. 33(6):420-7).Immunocytochemistry Analysis: A representative lot detected Klotho in Immunocytochemistry applications (Maltare, A., et. al. (2014). Monoclon Atib Immunodiagn Immunother. 33(6):420-7). | |
| EC: 3.2.1.31;KL | |
| Rat | |
| Unpurified | |
| RUO | |
| Primary | |
| Clone KL-234 is a rat monoclonal antibody that specifically detects Klotho. It targets an epitope with in 107 amino acids from the KL1 domain from the N-terminal region. | |
| Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
| IgG2a κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            .jpg-250.jpg)