Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRA1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356883 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356883 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356883 Supplier Novus Biologicals Supplier No. NBP176983PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLRA1 Polyclonal antibody specifically detects KLRA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen KLRA1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias killer cell lectin-like receptor subfamily A pseudogene 1, KLRA#, KLRA1, Ly49, Ly-49L, LY49L, MGC126520, MGC126522
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of human KLRA1. The immunogen is located within amino acids 190 - 240 of KLRA1. Amino Acid Squence: IGLSYDERESKWK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10748
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.