Learn More
Description
Specifications
Specifications
| Antigen | KLRC1/2/3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 20d5 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry |
| Gene Alias | CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B |
| Gene Symbols | KLRC1 |
| Host Species | Rat |
| Immunogen | CHO cells transfected with B6 allele of NKG2A gene |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
