Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRC1/2/3 Antibody (20d5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP128100X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128100X 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128100X Supplier Novus Biologicals Supplier No. NBP128100AF488
Add to Cart
Add to Cart

Rat Monoclonal Antibody

KLRC1/2/3 Monoclonal specifically detects KLRC1/2/3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen KLRC1/2/3
Applications Flow Cytometry
Classification Monoclonal
Clone 20d5
Conjugate Alexa Fluor 488
Dilution Flow Cytometry
Gene Alias CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Gene Symbols KLRC1
Host Species Rat
Immunogen CHO cells transfected with B6 allele of NKG2A gene
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3821
Test Specificity Mouse NKG2 (Mr approx. 43 kDa)
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.