Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRC4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131161 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131161 Supplier Novus Biologicals Supplier No. NBP131161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLRC4 Polyclonal specifically detects KLRC4 in Human samples. It is validated for Western Blot.

Specifications

Antigen KLRC4
Applications Western Blot
Classification Polyclonal
Concentration 0.15mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias D12S2489E, FLJ17759, killer cell lectin-like receptor subfamily C, member 4, natual killer cell group 2-F, NK cell receptor F, NKG2-F, NKG2-F type II integral membrane protein, NKG2-F-activating NK receptor, NKG2FFLJ78582
Gene Symbols KLRC4
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human KLRC4. The exact sequence is proprietary.
Molecular Weight of Antigen 18 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8302
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%). Not yet tested in any other species.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.