Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klre-1 Antibody (854929), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346688 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346688 Supplier Novus Biologicals Supplier No. FAB7099MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Klre-1 Monoclonal specifically detects Klre-1 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Klre-1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 854929
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD94
Host Species Rat
Immunogen Chinese hamster ovary cell line, CHO derived recombinant mouse Klre-1, Lys94-Lys226, Accession # NP_705818
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3824
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.