Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG1 Antibody (2388C), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311391 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311391 Supplier Novus Biologicals Supplier No. FAB70293PEATT594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

KLRG1 Monoclonal specifically detects KLRG1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KLRG1
Applications Flow Cytometry
Classification Monoclonal
Clone 2388C
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias 2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human KLRG1, Leu60-Phe195, Accession # Q96E93
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10219
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.