Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG1 Antibody (2F1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP128115X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128115X 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128115X Supplier Novus Biologicals Supplier No. NBP128115AF488
Add to Cart
Add to Cart

Golden Syrian Hamster Monoclonal Antibody

KLRG1 Monoclonal specifically detects KLRG1 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen KLRG1
Applications Flow Cytometry
Classification Monoclonal
Clone 2F1
Conjugate Alexa Fluor 488
Dilution Flow Cytometry
Gene Alias 2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
Gene Symbols KLRG1
Host Species Golden Syrian Hamster
Immunogen IL-2-activated NK cells (A-LAK cells) from B6 (H-2b) mice
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10219
Test Specificity KLRG1 (Mr 30-38 kDa)
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.