Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KMT2A/MLL Antibody, Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N60256APCY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N60256APCY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N60256APCY7 Supplier Novus Biologicals Supplier No. NB600256APCCY7
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KMT2A/MLL Polyclonal specifically detects KMT2A/MLL in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KMT2A/MLL
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALL1, ALL-1MLL/GAS7, CXXC7TET1-MLL, CXXC-type zinc finger protein 7, EC 2.1.1.43, HRXFLJ11783, HTRX, HTRX1MLL-AF4 der(11) fusion protein, KMT2ACDK6/MLL fusion protein, Lysine N-methyltransferase 2A, MLL/GAS7 fusion protein, MLL/GMPS fusion protein, MLL1, MLL1A, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog), myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila), Trithorax-like protein, TRX1histone-lysine N-methyltransferase MLL, Zinc finger protein HRX
Gene Symbols KMT2A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 2725 and 2775 of human myeloid/lymphoid or mixed-lineage leukemia 1 using the number given in Swiss-Prot entry Q03164 (GeneID 4297).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4297.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.