Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KNT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | KNT1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119465
|
Invitrogen™
PA5119465 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5122191
|
Invitrogen™
PA5122191 |
20 μL |
Each for $237.00
|
|
|||||
Description
KNT1 Polyclonal Antibody for Western Blot, IHC (P)
KNT1 (T-Kinninogen 1) is the acute phase kininogen of the rat. KNT1 could be a healing protein because of its properties as a cysteine protease inhibitor. Evidence has been presented that mRNA of T-kininogen synthesis may be a function of interleukin 6 production. In rats, four types of kininogens are produced, two of which are classical high and low molecular weight kininogens and two of which are low molecular weight-like kininogens, designated T-kininogen 1 and T-kininogen 2. T-kininogen 1 and T-kininogen 2 are 430 amino acid secreted rat proteins that each contain three cystatin domains and have nearly identical functions. Existing in plasma, both T-kininogen 1 and T-kininogen 2 are glycoproteins that act as thiol protease inhibitors and also play a role in blood coagulation, specifically by helping to optimally position blood coagulation factors.Specifications
KNT1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
Kng1 | |
T-Kinninogen 1 | |
Kng1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P08934 | |
288001 | |
Recombinant protein T-Kininogen 1. The antigen corresponds to amino acid range 378-430 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title