Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KNTC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10088131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10088131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10088131 Supplier Novus Biologicals Supplier No. NB10088131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KNTC1 Polyclonal specifically detects KNTC1 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen KNTC1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055523.1
Gene Alias FLJ36151, hRod, hsROD, KIAA0166Rough Deal homolog, centromere/kinetochore protein, kinetochore associated 1, kinetochore-associated protein 1, Rod, RODHsROD, Rough deal homolog
Gene Symbols KNTC1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2159 and 2209 of human rough deal homolog using the numbering given in entry NP_055523.1 (GeneID 9735).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9735
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.