Learn More
DWK Life Sciences Kimble™ Kontes™ Drying Tube for Microflex Kit
Shop All DWK Life Sciences Products
Click to view available options
:
Drying tube
Specifications
Specifications
For Use With (Equipment) | Microflex kits |
Product Type | Drying Tube |
Standard Taper Joint No. | 14/10 |