Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Liebig Condensers

Standard Taper outer joint at the top; standard Taper inner drip joint at the bottom.
Supplier: DWK Life Sciences 4470002430
Description
- Hose connections accept 3/8 in. ID flexible tubing
- Ref: ASTM Method D322
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Specifications
17.4 in. | |
24/40 | |
Borosilicate Glass | |
442 mm |
11.81 in. | |
USP Type I and ASTM E438, Type I, Class A requirements | |
Condenser | |
300 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction