Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KOR Antibody, Novus Biologicals™
SDP

Catalog No. NBP168860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP168860 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP168860 Supplier Novus Biologicals Supplier No. NBP168860
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

KOR Polyclonal specifically detects KOR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen KOR
Applications Western Blot, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P41145
Gene Alias kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK
Gene Symbols OPRK1
Host Species Goat
Immunogen Peptide with sequence C-PAYLRDIDGMNKP corresponding to C-Terminus according to NP_000903.2.
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4986
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.