Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kos1/Tnk1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB362220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362220 Supplier Novus Biologicals Supplier No. NB300237PCP

Rabbit Polyclonal Antibody

Kos1/Tnk1 Polyclonal antibody specifically detects Kos1/Tnk1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Kos1/Tnk1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias CD38 negative kinase 1;Kinase of embryonic stem cells;MGC46193;non-receptor tyrosine kinase;Non-receptor tyrosine-protein kinase TNK1;TNK1;tyrosine kinase, non-receptor 1;tyrosine kinase, non-receptor, 1
Host Species Rabbit
Immunogen C-terminal residues 421-434 [QNQVQSLPAGSAEP] of the 47 kDa mouse Kos1/ Tnk1 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.