Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOX8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179392
Description
KOX8 Polyclonal specifically detects KOX8 in Human samples. It is validated for Western Blot.Specifications
| KOX8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZp686L10267, FLJ16755, FLJ46310, KOX8, zinc finger protein 15, zinc finger protein 15-like 1 (KOX 8), zinc finger protein 708, zinc finger protein KOX8, ZNF15, ZNF15L1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7562 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| EAW84891 | |
| ZNF708 | |
| Synthetic peptide directed towards the C terminal of human ZNF708. Peptide sequence ILTKHKVIHTEDKPYKCEECGKTFNYSSNFTNHKKIHTGEKPYKCEECGK. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 100%; Human: 100%; Fruit fly: 90%; Rat: 90%; Crab-eating macaque: 78%; Chimpanzee: 76%; Sumatran orangutan: 76%; Bovine: 76%; African malaria mosquito: 76%. | |
| Human, Bovine, Equine, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction