Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOX8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
| Antigen | KOX8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
KOX8 Polyclonal specifically detects KOX8 in Human samples. It is validated for Western Blot.Specifications
| KOX8 | |
| Polyclonal | |
| Rabbit | |
| EAW84891 | |
| 7562 | |
| Synthetic peptide directed towards the C terminal of human ZNF708. Peptide sequence ILTKHKVIHTEDKPYKCEECGKTFNYSSNFTNHKKIHTGEKPYKCEECGK. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZp686L10267, FLJ16755, FLJ46310, KOX8, zinc finger protein 15, zinc finger protein 15-like 1 (KOX 8), zinc finger protein 708, zinc finger protein KOX8, ZNF15, ZNF15L1 | |
| ZNF708 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title