Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KPNA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166092 20 μg
NB166091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166092 Supplier Novus Biologicals Supplier No. NBP31549820UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KPNA3 Polyclonal antibody specifically detects KPNA3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen KPNA3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias hSRP1, importin alpha 4, Importin alpha Q2, importin alpha-3, importin subunit alpha-3, importin-alpha-Q2, IPOA4, karyopherin alpha 3 (importin alpha 4), Karyopherin subunit alpha-3, QIP2, SRP1, SRP1gamma, SRP1-gamma, SRP4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human KPNA3 (NP_002258.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3839
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.