Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KPNA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$149.90 - $405.59
Specifications
| Antigen | KPNA3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
KPNA3 Polyclonal antibody specifically detects KPNA3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| KPNA3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hSRP1, importin alpha 4, Importin alpha Q2, importin alpha-3, importin subunit alpha-3, importin-alpha-Q2, IPOA4, karyopherin alpha 3 (importin alpha 4), Karyopherin subunit alpha-3, QIP2, SRP1, SRP1gamma, SRP1-gamma, SRP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human KPNA3 (NP_002258.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 3839 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title