Learn More
Krackeler Scientific GC Inlet Liners
SDP
Catalog No. p-3206739
Click to view available options
Product Type:
Split Liner
Splitless Liner
Description:
Single Taper, Deactivated
Single Taper, Glass Wool, Deactivated
Straight, Glass Wool
Specifications
Description Straight, Glass Wool
Volume (Metric) 990 μL
Includes Glass wool
Product Type Split Liner
Diameter (Metric) Inner 4 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.