Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRAS Antibody (OTI2C1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289112 Supplier Novus Biologicals Supplier No. NBP271079APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KRAS Monoclonal antibody specifically detects KRAS in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot

Specifications

Antigen KRAS
Applications Western Blot
Classification Monoclonal
Clone OTI2C1
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias cellular c-Ki-ras2 proto-oncogene, c-Ki-ras, C-K-RAS, GTPase KRas, Ki-Ras, Kirsten rat sarcoma-2 viral (v-Ki-ras2) oncogene homolog, K-Ras 2, K-ras p21 protein, KRAS1, K-RAS2A, K-RAS2B, KRAS2NS3, K-RAS4A, K-RAS4B, NS, oncogene KRAS2, PR310 c-K-ras oncogene, RASK2c-Kirsten-ras protein, transforming protein p21, v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog, v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human KRAS (NP_203524) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3845
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.