Learn More
Thermo Scientific Chemicals Krebs-Heneseleit buffer solution
Click to view available options
Quantity:
500 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Plastic Bottle |
Frequently Asked Questions (FAQs)
What are the concentrations of the components of Krebs-Heneseleit buffer solution?
The Krebs-Heneseleit buffer solution contains:
- 118 mM sodium chloride
- 4.7 mM potassium chloride
- 1.25 mM calcium chloride
- 1.20 mM magnesium sulfate
- 1.20 mM monopotassium phosphate
- 25 mM sodium bicarbonate
- 10 mM HEPES
- 11.1 mM D-glucose