Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kremen-1 Antibody (RM0120-6K45), Biotin, Novus Biologicals™
SDP

Catalog No. NBP122498B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122498B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122498B Supplier Novus Biologicals Supplier No. NBP122498B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Kremen-1 Monoclonal specifically detects Kremen-1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Kremen-1
Applications Western Blot
Classification Monoclonal
Clone RM0120-6K45
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1Dickkopf receptor
Gene Symbols KREMEN1
Host Species Rat
Immunogen Mouse recombinant protein of the extracellular domain of Kremen-1
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83999
Test Specificity This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of Kremen-1.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.