Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kremen-2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB355715 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355715 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355715 Supplier Novus Biologicals Supplier No. NBP222339MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kremen-2 Polyclonal antibody specifically detects Kremen-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Kremen-2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Dickkopf receptor 2, kremen protein 2, kringle containing transmembrane protein 2, Kringle domain-containing transmembrane protein 2, KRM2Kringle-containing protein marking the eye and the nose, MGC10791, MGC16709
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human Kremen-2. The immunogen is located within amino acids 390-440 of Kremen-2. Amino Acid Sequence: ALGASRGGPRRSWAV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79412
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.