Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589501
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA589501 100 μL
1 options

Catalog No. PIPA589501

Supplier: Invitrogen™ PA589501

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYYGAVEEEK PQFEEEEGLE DDWNWDTWDG PEQEGDWSQQ ELHCEDPNFN MDADYDPSQP RKKKREAPLT GKKKRKSPFA AAVGQEKPVF EPGDKTFEEY LDEYYRLDYE DIIDDLPCRF KYRTVVPCDF GLSTEEILAA DDKELNRWCS LKKTCMYRSE QEELRDKRAY SQKAQNSWKK RQVFKSLCRE EAETPAEATG KPQRDEAGPQ RQLPALDGSL MGPESPPAQE EEAPVSPHKK PAPQKRRRAK KARLLGPTVM LGGCEFSRQR LQAFGLNPKR LHFRQLGRQR RKQQGPKNSS; Positive Samples: Jurkat.

This gene overlaps with the gene for cysteine endopeptidase AUT-like 4 in a head-to-tail orientation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRI1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KRI1
Gene Accession No. Q8N9T8
Gene Alias KRI1; KRI1 homolog; KRI1 homolog (S. cerevisiae); protein KRI1 homolog
Gene Symbols KRI1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 404-703 of human KRI1 (NP_075384.4).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65095
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.