Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRIT1 Rabbit anti-Human, Mouse, Clone: S01-6K9, Novus Biologicals™
SDP

Catalog No. NB188459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188459 25 μg
NB188458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188459 Supplier Novus Biologicals Supplier No. NBP31505325UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KRIT1 Monoclonal antibody specifically detects KRIT1 in Human, Mouse samples. It is validated for Western Blot, Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence

Specifications

Antigen KRIT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence
Classification Monoclonal
Clone S01-6K9
Conjugate Unconjugated
Dilution Western Blot, Simple Western, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunohistochemistry-Frozen, Immunofluorescence 20 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias CAMcerebral cavernous malformations 1, CCM1ankyrin repeat-containing protein Krit1, Cerebral cavernous malformations 1 protein, Krev interaction trapped 1, krev interaction trapped protein 1, KRIT1, ankyrin repeat containing
Host Species Rabbit
Immunogen Recombinant protein of human KRIT1 (Uniprot#: O00522)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 889
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.