Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRIT1 Rabbit anti-Human, Mouse, Clone: S01-6K9, Novus Biologicals™

Rabbit Monoclonal Antibody
$264.00 - $573.00
Specifications
Antigen | KRIT1 |
---|---|
Clone | S01-6K9 |
Dilution | Western Blot, Simple Western, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunohistochemistry-Frozen, Immunofluorescence 20 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence |
Classification | Monoclonal |
Description
KRIT1 Monoclonal antibody specifically detects KRIT1 in Human, Mouse samples. It is validated for Western Blot, Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ImmunofluorescenceSpecifications
KRIT1 | |
Western Blot, Simple Western, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunohistochemistry-Frozen, Immunofluorescence 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CAMcerebral cavernous malformations 1, CCM1ankyrin repeat-containing protein Krit1, Cerebral cavernous malformations 1 protein, Krev interaction trapped 1, krev interaction trapped protein 1, KRIT1, ankyrin repeat containing | |
Recombinant protein of human KRIT1 (Uniprot#: O00522) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S01-6K9 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
889 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title