Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRR1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP222252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222252 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222252 Supplier Novus Biologicals Supplier No. NBP222252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRR1 Polyclonal specifically detects KRR1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen KRR1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_008974.5
Gene Alias HIV-1 Rev binding protein 2, HIV-1 Rev-binding protein 2, HRB2, KRR1 small subunit processome component homolog, KRR1, small subunit (SSU) processome component, homolog (yeast), Rev interacting protein, Rev-interacting protein 1, Rip-1
Gene Symbols KRR1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 300 and 350 of human Small Subunit (SSU) Processome Component using the numbering given in entry NP_008974.5 (GeneID 11103).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11103
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.