Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT23 Antibody (OTI3F2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271780 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271780 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271780 Supplier Novus Biologicals Supplier No. NBP271780
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KRT23 Monoclonal specifically detects KRT23 in Human samples. It is validated for Western Blot.

Specifications

Antigen KRT23
Applications Western Blot
Classification Monoclonal
Clone OTI3F2
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias CK23, CK-23, cytokeratin 23, Cytokeratin-23, DKFZp434G032, HAIK1, hyperacetylation-inducible type I keratin, K23histone deacetylase inducible keratin 23, keratin 23 (histone deacetylase inducible), keratin, type I cytoskeletal 23, keratin-23, MGC26158, type I intermediate filament cytokeratin
Gene Symbols KRT23
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-270 of human KRT23(NP_056330) produced in E.coli.
Molecular Weight of Antigen 48 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 25984
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.