Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT24 Antibody (OTI4E8), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP2724A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2724A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2724A30 Supplier Novus Biologicals Supplier No. NBP272455AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KRT24 Monoclonal specifically detects KRT24 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen KRT24
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4E8
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CK-24, Cytokeratin-24, FLJ20261cytokeratin-24, K24, KA24, keratin 24, keratin, type I cytoskeletal 24, keratin-24, MGC138169, MGC138173, Type I keratin-24
Gene Symbols KRT24
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 139-397 of human KRT24 (NP_061889) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 192666.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.