Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT2B 293T Cell Overexpression Lysate (Denatured), Abnova

Catalog No. 89002223
Click to view available options
Quantity:
100 μL

Western Blotting

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description KRT76 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_015848.1
Molecular Weight (g/mol) 65.9
pH Range 6.8
Plasmid pCMV-KRT76 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-KRT76 antibody (H00051350-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.