Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT81 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KRT81 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KRT81 Polyclonal specifically detects KRT81 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KRT81 | |
Polyclonal | |
Rabbit | |
Human | |
ghHb1, ghHkb1, Hair keratin K2.9, hard keratin, type II, 1, Hb-1, hHAKB2-1, K81, keratin 81, Keratin, hair, basic, 1MLN 137, keratin, type II cuticular Hb1, Keratin-81, KRTHB1HB1, Metastatic lymph node 137 gene protein, MLN137, Type II hair keratin Hb1, Type-II keratin Kb21 | |
KRT81 | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
3887 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AESWYRSKCEEMKATVIRHGETLRRTKEEINELN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title