Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRTAP1-5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179847
Description
KRTAP1-5 Polyclonal specifically detects KRTAP1-5 in Human samples. It is validated for Western Blot.Specifications
KRTAP1-5 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
KAP1.5High sulfur keratin-associated protein 1.5, keratin associated protein 1.5, keratin associated protein 1-5, Keratin-associated protein 1.5, keratin-associated protein 1-5, KRTAP1.5, KRTAP-15, MGC126604 | |
Rabbit | |
18 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Mouse: 86%. | |
Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Goat, Rabbit, Sheep | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_114163 | |
KRTAP1-5 | |
Synthetic peptide directed towards the C terminal of human KRTAP1-5The immunogen for this antibody is KRTAP1-5. Peptide sequence TGCGIGGGISYGQEGSSGAVSTRIRWCRPDSRVEGTYLPPCCVVSCTPPS. | |
Affinity purified | |
RUO | |
83895 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction