Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRTAP2-4 Antibody (OTI3B4), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N272304J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272304J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272304J646 Supplier Novus Biologicals Supplier No. NBP272304JF646

Mouse Monoclonal Antibody

KRTAP2-4 Monoclonal specifically detects KRTAP2-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen KRTAP2-4
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3B4
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias High sulfur keratin-associated protein 2.4, KAP2.4MGC119884, keratin associated protein 2-4, Keratin-associated protein 2.4, keratin-associated protein 2-4, KRTAP2.4, MGC119882, MGC119885, MGC74790
Gene Symbols KRTAP2-4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-128 of human KRTAP2-4 (NP_149440) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 85294.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.