Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KSHV ORF26 Antibody (2F6B8), Novus Biologicals™
SDP

Catalog No. NBP147357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147357 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147357 Supplier Novus Biologicals Supplier No. NBP147357
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KSHV ORF26 Monoclonal antibody specifically detects KSHV ORF26 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen KSHV ORF26
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F6B8
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Gene Alias HHV8, Human Herpes Virus 8, Kaposi's sarcoma herpes virus, KSHV, ORF45
Gene Symbols ORF26
Host Species Mouse
Immunogen Purified recombinant fragment of KSHV ORF26 expressed in E. Coli.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4961509
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.