Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KSHV ORF57 Antibody, Novus Biologicals™
SDP

Catalog No. NB024680 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024680 50 μg
NB024679 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB024680 Supplier Novus Biologicals Supplier No. NBP29526050UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KSHV ORF57 Polyclonal specifically detects KSHV ORF57 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen KSHV ORF57
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1.55 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:8000, ELISA 1:100000 - 170000, Immunohistochemistry
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Alias herpesvirus core gene UL54 family, ORF57
Host Species Rabbit
Immunogen This KSHV ORF57 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of human KSHV ORF57 protein (Uniprot #: Q2HR75).
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4961525
Target Species Virus
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.