Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KSHV ORF57 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$366.00 - $670.50
Specifications
Antigen | KSHV ORF57 |
---|---|
Concentration | 1.55 mg/ml |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KSHV ORF57 Polyclonal specifically detects KSHV ORF57 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
KSHV ORF57 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Virus | |
herpesvirus core gene UL54 family, ORF57 | |
IgG | |
Immunogen affinity purified |
1.55 mg/ml | |
Polyclonal | |
Purified | |
RUO | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
4961525 | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title