Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KT 5720
GREENER_CHOICESDP

Catalog No. 1288100U Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-881-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-881-00U Supplier Tocris Bioscience™ Supplier No. 1288/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective protein kinase A inhibitor

Potent, selective inhibitor of protein kinase A (Ki = 60 nM). Has no effect on PKG or PKC (Ki > 2 μM). Reversibly arrests human skin fibroblasts in the G1 phase.

Chemical Identifiers

CAS 108068-98-0
Molecular Formula C32H31N3O5

Specifications

Quantity 100 μg
Formula Weight 537.61
Purity >98%
Molecular Weight (g/mol) 537.62
Chemical Name or Material KT 5720
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.