Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KU 55933
GREENER_CHOICESDP

Catalog No. 354410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-441-0 Supplier Tocris Bioscience™ Supplier No. 3544/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ATM kinase inhibitor

Potent, selective and competitive ATM kinase inhibitor (Ki = 2.2 nM, IC50 values are 13, 2500, 9300, 16600, > 100000 and > 100000 nM at ATM, DNA-PK, mTOR, PI 3-kinase, PI 4-K and ATR respectively). Decreases viability of MCF-7, A549 and HCT116 cells and decreases p21CIP1 levels in vitro. Acts as a radio- and chemosensitizer for the treatment of cancer.

Chemical Identifiers

CAS 587871-26-9
Molecular Formula C21H17NO3S2

Specifications

Quantity 10 mg
Formula Weight 395.49
Purity >98%
Molecular Weight (g/mol) 395.49
Chemical Name or Material KU 55933
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.