Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KU70/XRCC6 Mouse anti-Human, Mouse, Rat, Clone: 3C12D9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | KU70/XRCC6 |
---|---|
Clone | 3C12D9 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7025
![]() ![]() |
Proteintech Group Inc
666071IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.Specifications
KU70/XRCC6 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
5 dRP lyase Ku70, 70 kDa subunit of Ku antigen, CTC75, CTCBF, DNA repair protein XRCC6, G22P1, KU70, ML8, Thyroid lupus autoantigen, TLAA, XRCC6 | |
Xrcc6 | |
IgG1 | |
Protein G | |
Antibody |
3C12D9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P12956, P23475 | |
14375, 25019, 2547 | |
KU70,XRCC6 Fusion Protein Ag21301 | |
Primary | |
-20°C | |
XRCC6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title