Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ku70/XRCC6 Mouse, Clone: KU729, Novus Biologicals™
SDP

Catalog No. NBP234247A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234247A 0.1 mg
NBP234247F 0.02 mg
NBP234247B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP234247A Supplier Novus Biologicals Supplier No. NBP2342470.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ku70/XRCC6 Monoclonal specifically detects Ku70/XRCC6 in Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).

Specifications

Antigen Ku70/XRCC6
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone KU729
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P12956
Gene Alias 5'-deoxyribose-5-phosphate lyase Ku70, 5'-dRP lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S731, EC 3.6.4.-, EC 4.2.99.-, G22P1Ku70, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa, KU70, ML8,70 kDa subunit, Thyroid-lupus autoantigen, thyroid-lupus autoantigen p70, TLAAD22S671, X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6, X-ray repair cross-complementing protein 6
Gene Symbols XRCC6
Host Species Mouse
Immunogen Nuclear extract of human HL-60 cells
Molecular Weight of Antigen 75 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining
Primary or Secondary Primary
Gene ID (Entrez) 2547
Test Specificity Recognizes a dimer of two proteins of 70kDa and ∼80kDa, identified as two subunits of Ku. This MAb recognizes a conformational epitope of p70/p80 dimer, which is destroyed during Western blotting. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.
Target Species Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.