Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ku80/XRCC5 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345100 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345100 Supplier Novus Biologicals Supplier No. NB100508MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ku80/XRCC5 Polyclonal specifically detects Ku80/XRCC5 in Hamster, Human, Mouse, Rat samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.

Specifications

Antigen Ku80/XRCC5
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias 80kDa, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC85, CTCBF, EC 3.6.4.-, FLJ39089, G22P2, KARP1, KARP-1, KU80, Ku86 autoantigen related protein 1,86 kDa subunit of Ku antigen, Ku86DNA repair protein XRCC5, KUB2, NFIV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining)CTC box-binding factor 85 kDa subunit, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining; Ku autoantigen, 80kD), X-ray repair cross-complementing protein 5, X-ray repair, complementing defective, repair in Chinese hamster
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues 419-440 LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7520
Target Species Hamster, Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.