Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv11.2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | Kv11.2 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Kv11.2 Polyclonal antibody specifically detects Kv11.2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Kv11.2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, pH 7.2, 40% glycerol | |
81033 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
eag related protein 2, eag-related gene member 2, Eag-related protein 2, erg2, ERG-2, ether-a-go-go related gene potassium channel 2, Ether-a-go-go-related gene potassium channel 2, Ether-a-go-go-related protein 2, hERG-2, HERG2, Kv11.2, potassium voltage-gated channel subfamily H member 6, potassium voltage-gated channel, subfamily H (eag-related), member 6, Voltage-gated potassium channel subunit Kv11.2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SGSPHELGPQFPSKGYSLLGPGSQNSMGAGPCAPGHPDAAPPLSISDASGLWPELLQEMPPRHSPQSPQED | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title