Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv12.1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018491 0.025 mL
NBP130646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018491 Supplier Novus Biologicals Supplier No. NBP1306460.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kv12.1 Polyclonal specifically detects Kv12.1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Kv12.1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Accession No. Q96L42
Gene Alias ELK, ELK channel 3, ELK1, ELK3, ether-a-go-go-like potassium channel 1, Ether-a-go-go-like potassium channel 3, hElk1, Kv12.1, potassium voltage-gated channel subfamily H member 8, potassium voltage-gated channel, subfamily H (eag-related), member 8, Voltage-gated potassium channel subunit Kv12.1
Gene Symbols KCNH8
Host Species Rabbit
Immunogen A synthetic peptide from the c-terminal of human KCNH8 (Potassium voltage-gated channel subfamily H member 8) conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131096
Test Specificity KCNH8.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.