Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv12.1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$149.33 - $480.74

Specifications

Antigen Kv12.1
Concentration LYOPH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018491
SDP
View Documents
Novus Biologicals
NBP1306460.025ML
0.025 mL
Each for $149.33
Only null left
Add to Cart
 
NBP130646
SDP
View Documents
Novus Biologicals
NBP130646
0.1 mL
Each for $480.74
Only null left
Add to Cart
 
Description

Description

Kv12.1 Polyclonal specifically detects Kv12.1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Kv12.1
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Human
ELK, ELK channel 3, ELK1, ELK3, ether-a-go-go-like potassium channel 1, Ether-a-go-go-like potassium channel 3, hElk1, Kv12.1, potassium voltage-gated channel subfamily H member 8, potassium voltage-gated channel, subfamily H (eag-related), member 8, Voltage-gated potassium channel subunit Kv12.1
KCNH8
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
LYOPH
Polyclonal
Antisera
RUO
Q96L42
131096
IgG
Unpurified
KCNH8.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.