Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv3.2 Antibody (S410-17), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB259328700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259328700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259328700 Supplier Novus Biologicals Supplier No. NBP259328AF700

Mouse Monoclonal Antibody

Kv3.2 Monoclonal specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Kv3.2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S410-17
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2
Gene Symbols KCNC2
Host Species Mouse
Immunogen Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3747
Test Specificity Detects 80-100kDa. Varies due to post-translational modifications.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.