Learn More
Description
Specifications
Specifications
| Antigen | Kv3.3 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | KSHIIID, KV3.3, potassium voltage-gated channel subfamily C member 3, potassium voltage-gated channel, Shaw-related subfamily, member 3, SCA13, Shaw-related voltage-gated potassium channel protein 3, spinocerebellar ataxia 13, voltage-gated potassium channel protein KV3.3, Voltage-gated potassium channel subunit Kv3.3 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human Kv3.3 (NP_004968). Peptide sequence YAERIGADPDDILGSNHTYFKNIPIGFWWAVVTMTTLGYGDMYPKTWSGM |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
