Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV4.1 (KCND1) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595216
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595216 Supplier Invitrogen™ Supplier No. PA595216
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human KCND1 shares 90% amino acid (aa) sequence identity with mouse KCND1. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Mouse Brain Tissue, HELA whole cell, COLO320 whole cell, A549 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The KV gene family encodes more than 30 genes that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha-subunits (KV alpha), which include the KV1, KV2, KV3 and KV4 proteins, and accessory or KV beta subunits that modify the gating properties of the coexpressed KV alpha subunits. Differences exist in the patterns of trafficking, biosynthetic processing and surface expression of the major KV1 subunits (KV1. 1, KV1. 2 and KV1. 4) expressed in rat and human brain, suggesting that the individual protein subunits are highly regulated to control for the assembly and formation of functional neuronal channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV4.1 (KCND1)
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Kcnd1
Gene Accession No. Q03719, Q9NSA2
Gene Alias 1110037K09Rik; Kca2-1; Kcnd1; KV4.1; mShal; mShal1; potassium channel, voltage gated Shal related subfamily D, member 1; potassium channel, voltage gated Shal-related subfamily D, member 1; potassium voltage gated channel Shal-related family member 1; potassium voltage gated channel, Shal-related family, member 1; potassium voltage-gated channel subfamily D member 1; potassium voltage-gated channel, Shal-related family, member 1; potassium voltage-gated channel, Shal-related subfamily, member 1; Shal; Shal1; Shal-type potassium channel; Voltage-gated potassium channel subunit Kv4.1
Gene Symbols Kcnd1
Host Species Rabbit
Immunogen E.coli-derived human KCND1 recombinant protein (Position: T442-L647).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16506, 3750
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.